SYNS181530
Synthware™ straight bore PTFE stopcock plug
bore size 4 mm, Plug Size: 15.2/30
Synonym(s):
Synthware S181530
All Photos(1)
About This Item
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
Synthware S181530
bore size
4 mm
Related Categories
General description
This replacement kit is complete with one PTFE plug (1:5 taper), one PTFE washer, one O-ring.
Legal Information
Synthware is a trademark of Kemtech America Inc.