Key Documents
SYND151819
Synthware short path distillation head
joint: ST/NS 19/22
Synonym(s):
Synthware D151819, Short path head
Select a Size
1 EA
CHF 188.00
CHF 188.00
Check Cart for Availability
Select a Size
Change View
1 EA
CHF 188.00
About This Item
UNSPSC Code:
41104812
NACRES:
NB.51
CHF 188.00
Check Cart for Availability
Joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware D151819
Related Categories
General description
Short-path design for microscale application, used with 25 mm immersion thermometer, and D27 distillation receiver.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)