Key Documents
SYNA852440
Synthware U-shaped drying tube
joint: ST/NS 24/40
Synonym(s):
Synthware A852440, Drying tube adapter
Select a Size
1 EA
CHF 50.60
CHF 50.60
Check Cart for Availability
Select a Size
Change View
1 EA
CHF 50.60
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
CHF 50.60
Check Cart for Availability
material
colorless tube
glass tube
Joint
joint: ST/NS24/40
sterility
non-sterile
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A852440
General description
With joint. Cotton is usually used to contain drying agent.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)