Documents
SYNA491420
Synthware vacuum distilling adapter
joint: ST/NS 14/20, hose size 8 mm, Distance Between Joints: 100 mm
Synonym(s):
Synthware A491420, glass distilling adapter
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Joint
joint: ST/NS14/20
packaging
pack of 1 ea
manufacturer/tradename
A491420
hose size
8 mm
Related Categories
Legal Information
Synthware is a trademark of Kemtech America Inc.