Skip to Content
Merck
All Photos(1)

Key Documents

SLW4536/32D

Pyrex® High temperature pouring ring

thread fits 32 mm

Synonym(s):

Pyrex, SciLabware 4536/32D

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
CHF 23.80

CHF 23.80


Check Cart for Availability


Select a Size

Change View
10 EA
CHF 23.80

About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

CHF 23.80


Check Cart for Availability

material

red ETFE

manufacturer/tradename

4536/32D
SciLabware

fitting

thread fits 32 mm

Looking for similar products? Visit Product Comparison Guide

General description

  • For use with 1516, 1517, 1518, 1519 and 1520 series bottles
  • Ideal for applications involving high temperatures
  • Red coloured ethylene-tetrafluoroethylene pouring ring
  • Withstands hot air sterilisation up to 180 deg. C

Legal Information

PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service