Skip to Content
Merck
All Photos(1)

Documents

SLW4500/02M

Pyrex® Square plastic feet

for 50 mL measuring cylinder

Synonym(s):

Pyrex, SciLabware 4500/02M

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

material

polyethylene

manufacturer/tradename

4500/02M
SciLabware

Looking for similar products? Visit Product Comparison Guide

General description

For use with SLW3205 cylinders

Features and Benefits

• Manufactured from polyethylene• Replacement feet for PYREX® 3205 range of measuring cylinders.• Hexagonal foot shape

Legal Information

PYREX is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service