Skip to Content
Merck
All Photos(1)

Key Documents

SLW1597/04D

Pyrex® Desiccator plate

plate size 200 mm

Synonym(s):

Pyrex, SciLabware 1597/04D

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CHF 74.70

CHF 74.70


Check Cart for Availability


Select a Size

Change View
1 EA
CHF 74.70

About This Item

UNSPSC Code:
41121800
NACRES:
NB.43
material:
metal

CHF 74.70


Check Cart for Availability

material

metal

manufacturer/tradename

1597/04D
SciLabware

plate size

200 mm

Looking for similar products? Visit Product Comparison Guide

General description

Metal perforated plate for use with Pyrex vacuum and non-vacuum desiccators

Features and Benefits

  • Manufactured from metal
  • Plate perforated
  • For use with PYREX® vacuum and non-vacuum desiccators

Legal Information

PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service