Skip to Content
Merck
All Photos(1)

Key Documents

SLW1571/02M

Pyrex® Aspirator stopcock with ground cone

bore size 3.0 mm, core joint: 29/32

Synonym(s):

Pyrex, SciLabware 1571/02M

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CHF 97.60

CHF 97.60


Check Cart for Availability


Select a Size

Change View
1 EA
CHF 97.60

About This Item

UNSPSC Code:
41121500
NACRES:
NB.43

CHF 97.60


Check Cart for Availability

Joint

joint: core29/32

manufacturer/tradename

1571/02M
SciLabware

bore size

3.0 mm

General description

  • For use with Aspirator bottles 1541 and 1569
  • With traditional glass key

Legal Information

PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service