Skip to Content
Merck
All Photos(1)

Key Documents

SLW1515/08D

Pyrex® Reagent bottles, round bottom with reusable screw caps

capacity 500 mL

Synonym(s):

Pyrex, SciLabware 1515/08D

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
CHF 130.00

CHF 130.00


Check Cart for Availability


Select a Size

Change View
10 EA
CHF 130.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

CHF 130.00


Check Cart for Availability

material

PTFE seal
Pyrex bottle
black PBT cap
colorless bottle
round bottle
round bottom
white graduation

Joint

Joints threaded neck (SVL 30)

feature

with cap

packaging

pack of 10 ea

manufacturer/tradename

SciLabware 1515/08D

bottle diam. × H

81 mm × 175 mm

cap size

30 mm

capacity

500 mL

Looking for similar products? Visit Product Comparison Guide

General description

  • Complies with ISO 4796-1
  • Ideal for storage of reagents, analytical standards and culture media etc.
  • Manufactured from Pyrex borosilicate glass
  • Excellent resistance to chemical attack and high temperatures
  • Supplied with PBT screwcaps having a PTFE faced sealing disc
  • Withstands hot air sterilisation up to 180 deg. C
  • Autoclavable
  • White graduations and marking spot

Legal Information

PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service