Key Documents
SLW1060/14D
Pyrex® Watch glasses
nominal diameter 100 mm
Synonym(s):
Pyrex, SciLabware 1060/14D
Select a Size
10 EA
CHF 234.00
CHF 234.00
Check Cart for Availability
Select a Size
Change View
10 EA
CHF 234.00
About This Item
UNSPSC Code:
41121801
NACRES:
NB.43
CHF 234.00
Check Cart for Availability
packaging
pkg of 10
manufacturer/tradename
1060/04D
SciLabware
nominal diameter
100 mm
General description
Watch glasses
  • For evaporation work and covering heated beakers
  • Manufactured from heat resisting Pyrex borosilicate glass
  • Fire-polished edges to reduce chipping and breakage
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)