SLW1015/22D
Pyrex® Berzelius beakers, graduated, tall form with spout, with printed trace code
1000 mL
Synonym(s):
Pyrex, SciLabware 1015/22D
All Photos(1)
About This Item
Recommended Products
feature
graduated
manufacturer/tradename
1015/22D
SciLabware
H
180 mm
O.D.
95 mm
capacity
1000 mL
General description
- Complies with ISO 3819 and DIN 12331
- Manufactured from Pyrex® borosilicate glass
- Printed with Trace Code for downloadable batch certificate
- Batch certificate available to download from www.scilabware.com
- Tall form with spout
- White graduations and marking spot
Other Notes
Browse all glass beakers
Legal Information
PYREX is a registered trademark of Corning, Inc.