Skip to Content
Merck
All Photos(1)

Key Documents

SIK7121-10

Sigma-Aldrich

12,12,13,13,14,14,15,15,16,16,17,17-Tridecafluoroseptadecylphosphonic acid

95%

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
CHF 147.00
1 G
CHF 442.00

CHF 147.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
CHF 147.00
1 G
CHF 442.00

About This Item

Empirical Formula (Hill Notation):
C17H24F13O3P
Molecular Weight:
554.32
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.23

CHF 147.00


Check Cart for Availability

Request a Bulk Order

Assay

95%

form

powder

shelf life

3 years under the recommended conditions

color

white

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCCCCCCCCCC[P](=O)(O)O

InChI key

BUBQNXNENZFVMX-UHFFFAOYSA-N

Application

These products are used to form water repellent and/or lubricated surfaces on metal oxide materials (TiO2, ZnO, etc.).
The fluorinated carbon gives the omniphobic properties to the surface. The coated surface is more water repellent when a product having more fluorinated carbons is used.
The phosphonic acids groups can covalently bond to metal oxide surfaces to form sufficient self-assembled monolayer.

Legal Information

Product of SiKEMIA

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service