Skip to Content
Merck
All Photos(1)

Key Documents

QBD10615

Sigma-Aldrich

Fmoc-N-Lys-(dPEG®12-biotin)-OH-(acid)

>95% (HPLC)

Synonym(s):

Fmoc-Lys(biotin-PEG12)-OH, Fmoc-N-Lys(PEG12-biotin)-OH-(acid), Fmoc-PEG-biotin acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 238.00

CHF 238.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
100 MG
CHF 238.00

About This Item

Empirical Formula (Hill Notation):
C58H91N5O19S
Molecular Weight:
1194.43
UNSPSC Code:
12352106
NACRES:
NA.22

CHF 238.00


Estimated to ship on01 June 2025


Request a Bulk Order

Assay

>95% (HPLC)

form

solid or viscous liquid

reaction suitability

reaction type: Biotinylations
reaction type: Pegylations

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

Features and Benefits

Fmoc-N-Lys-(dPEG12-biotin)-OH permits incorporation of a medium-length (40 atoms, 47.5 Å), hydrophilic, biotinylated discrete PEG (dPEG) into a peptide chain using standard Fmoc chemistry with Fmoc-protected lysine. The biotin-dPEG12 moiety is coupled to the side chain of lysine. A useful application for this product is the creation of a peptide probe that can be retrieved by affinity capture using avidin or streptavidin.

Automate your Biotin tagging with Synple Automated Synthesis Platform (SYNPLE-SC002)

Legal Information

Products Protected under U.S. Patent #s 7,888,536 & 8,637,711 and European Patent #s 1,594,440 & 2,750,681
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service