Key Documents
PL800GABUT-10
Plas-Labs replacement butyl gloves
for 2 piece glove set, size 10 (for 8 in. ports)
Select a Size
1 PAIR
CHF 196.00
CHF 196.00
Check Cart for Availability
Select a Size
Change View
1 PAIR
CHF 196.00
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
CHF 196.00
Check Cart for Availability
material
butyl rubber
size
10 (for 8 in. ports)
feature
reusable
for use with glove box sleeves
packaging
pack of 1 pair
manufacturer/tradename
Plas-Labs 800GABUT-10
General description
Plas-Labs black butyl rubber gloves for highest impermeability to water vapor, gases, and toxic fluids
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
Choose from one of the most recent versions:
Certificates of Analysis (COA)