Skip to Content
Merck
All Photos(1)

Key Documents

P67609

Sigma-Aldrich

4-Pyridylcarbinol N-oxide

98%

Synonym(s):

4-Pyridinemethanol N-oxide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CHF 63.60

CHF 63.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
CHF 63.60

About This Item

Empirical Formula (Hill Notation):
C6H7NO2
CAS Number:
Molecular Weight:
125.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

CHF 63.60


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

crystals

mp

124-127 °C (lit.)

SMILES string

OCc1cc[n+]([O-])cc1

InChI

1S/C6H7NO2/c8-5-6-1-3-7(9)4-2-6/h1-4,8H,5H2

InChI key

ZJGXPKKVSNSGRJ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service