Skip to Content
Merck
All Photos(1)

Documents

P55880

Sigma-Aldrich

Purin-6-yltrimethylammonium chloride

Synonym(s):

NSC-51095, Trimethylpurin-6-ylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12ClN5
CAS Number:
Molecular Weight:
213.67
MDL number:

SMILES string

[Cl-].C[N+](C)(C)c1ncnc2nc[nH]c12


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service