Skip to Content
Merck
All Photos(1)

Key Documents

N28000

Sigma-Aldrich

5-Nitrovanillin

97%

Synonym(s):

4-Hydroxy-3-methoxy-5-nitrobenzaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
CHF 44.40

CHF 44.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
CHF 44.40

About This Item

Linear Formula:
5-O2NC6H2-3-(OCH3)-4-(OH)CHO
CAS Number:
Molecular Weight:
197.14
Beilstein:
1973746
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

CHF 44.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder

mp

172-175 °C (lit.)

SMILES string

[H]C(=O)c1cc(OC)c(O)c(c1)[N+]([O-])=O

InChI

1S/C8H7NO5/c1-14-7-3-5(4-10)2-6(8(7)11)9(12)13/h2-4,11H,1H3

InChI key

ZEHYRTJBFMZHCY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

206.6 °F

Flash Point(C)

97 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service