Skip to Content
Merck
All Photos(1)

Key Documents

MT51350038

Mettler-Toledo certified buffer

pH 10, volume 250 mL

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
CHF 32.20

CHF 32.20


Check Cart for Availability


Select a Size

Change View
250 ML
CHF 32.20

About This Item

UNSPSC Code:
51191600
NACRES:
NB.79

CHF 32.20


Check Cart for Availability

sterility

non-sterile

form

liquid

manufacturer/tradename

Mettler-Toledo 51350038

volume

250 mL

pH

10

Looking for similar products? Visit Product Comparison Guide

General description

The perfect product for regulated industries. These solutions are traceable to a certificate of an accredited body of DKD.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service