Key Documents
MT51340012
Mettler Toledo DX series ISE half-cell membrane kit
For Ammonium ISE
Select a Size
1 EA
CHF 795.00
CHF 795.00
Check Cart for Availability
Select a Size
Change View
1 EA
CHF 795.00
About This Item
UNSPSC Code:
39121436
NACRES:
NB.79
CHF 795.00
Check Cart for Availability
manufacturer/tradename
Mettler-Toledo 51340012
General description
DX series ion specific membrane kit. Mount the new membrane onto the shaft of your ISE half-cell, and you have a new ISE!
Kit contains one membrane plus electrolyte
Choose from one of the most recent versions:
Certificates of Analysis (COA)