MT51302613
Mettler-Toledo SevenGo™ Duo pH/conductivity SG23
SG68 field kit, InLab® Expert Pro-ISM and InLab® 605-ISM, cable L 5 m
Select a Size
CHF 3’470.00
Select a Size
About This Item
CHF 3’470.00
Recommended Products
description
Calibration pH: Max. 5 points, 7 predefined buffer groups; DO: 1 or 2 points
DO range 0.0 ... 600%, 0.00 ... 99.00 mg/L (+/- 0.5%)
Temperature range °C pH ATC: -5.0 ... 105.0; MTC: -30 ... 130/DO: 0.0 ... 60.0 (+/- 0.2)
manufacturer/tradename
Mettler-Toledo 51302613
mV measuring range
-1999.9-1999.9, accuracy: ±0.1
cable L
5 m
battery power
4 AA (included in standard delivery)
pH range
-2-19.999
General description
The revolutionary intelligent sensor management system (ISM®) offers the user an even greater level of security. The calibration data is stored on the chip in the sensor and is used automatically when the sensor is connected to the meter. This ensures that the meter always uses the latest calibration data.
Features and Benefits
pH/Ion/Oxygen Meter (IP67) for the Strictest Demands
Portable dual-channel meter for the parallel determination of pH, mV, rel. mV or ion concentrations and dissolved oxygen (DO)
Either linear or segmented 5-point pH calibration with selection from 7 predefined and one user-defined buffer groups
Manual or automatic atmospheric pressure compensation using built-in barometer
Modern data management for 500 GLP data records with date/time, sensor ID and serial number, user and sample ID
Contactless infrared interface for data transmission to a printer or PC
Intuitive, user-friendly operation using menu guidance in 10 languages and backlit display
Rugged watertight housing (IP67)
Revolutionary intelligent sensor management (ISM)
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)