Key Documents
MT30040000
Mettler-Toledo uMix magnetic stirrer, for use with SevenCompact and SevenExcellence
Must be connected to meter
Select a Size
1 EA
CHF 407.00
CHF 407.00
Check Cart for Availability
Select a Size
Change View
1 EA
CHF 407.00
About This Item
UNSPSC Code:
41103800
NACRES:
NB.79
CHF 407.00
Check Cart for Availability
manufacturer/tradename
Mettler-Toledo 30040000
General description
The uMix magnetic stirrer from Mettler Toledo provides an extra degree of precision for your applications. Powered and controlled externally, the magnetic stirrer allows for enhanced reproducibility of results and higher efficiency in the measuring process.
The magnetic stirrer can be combined with either the SevenCompact pH/Ion S220, the SevenCompact Conductivity S230, or the multiparameter SevenExcellence benchtop meters.
Legal Information
SevenCompact is a trademark of Mettler-Toledo, Inc.
SevenExcellence is a trademark of Mettler-Toledo, Inc.
uMix is a trademark of Mettler-Toledo, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)