Skip to Content
Merck
All Photos(1)

Key Documents

MT30019034

Mettler-Toledo SevenCompact conductivity meters S230

conductivity S230 kit, AC/DC input 230 V AC, universal plug set

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115609
NACRES:
NB.61

description

Calibration 13 pre-defined and 1 user-defined standard;
Conductivity range 0.001 ?S/cm to 1000 mS/cm, autoscaling (+/-0.5%)
Resistivity range 0.00 ...100 MOhm*cm, autoscaling

feature

kit incl. InLab® 731-ISM, electrode holder and calibration sachets

manufacturer/tradename

Mettler-Toledo 30019034

TDS measuring range

0-1000 g/L

salinity measuring range

0-80 (psu)

Looking for similar products? Visit Product Comparison Guide

General description

Includes meter, electrode holder, InLab® 731-ISM electrode and calibration sachets.
Mettler Toledo SevenCompact conductivity meter S230 is the perfect choice for a wide range of applications without overstretching your budget. Not only does it measure conductivity but also various other parameters such as salinity, resistivity, total dissolved solids and conductivity ash.

The included electrode holder with the perfectly vertical uPlace movement helps you bring your electrode in the position where it works best for your sample. This allows for faster measurements and reduces the risk of the sample vessel tipping over and/or damage the sensor.

SevenCompact gives you the choice to view all measurement information on the display or to focus on the essentials with the uFocus view. Switching between the different display layouts is easy, requiring only a single keypress.

The new color display with its well-designed icons and menu settings in 10 languages makes operation genuinely intuitive. Applications range from routine measurements through sample analysis, data handling and data archiving in compliance with GLP regulations. Simply innovative design that meets the requirements for a universal, easy-to-operate conductivity meter.

  • User friendly instrument for demanding users
  • Security and high reproducibility thanks to Intelligent Sensor Management (ISM®)
  • Improved quality of measurements through professional calibration support
  • Comprehensive service package including IQ/OQ
  • Integrated USB and RS232 interfaces for data exchange

Features and Benefits

Prominent color display
The 4.3 inch high-resolution color display with its large digits and well arranged icons makes relevant information identifiable at a glance.

Two display layouts
Switch with a single keypress between the full-information screen or the uFocus layout, in which you will not be distracted by information of less importance.

IntelligentSensor Management (ISM)
ISM helps eliminate mistakes by automatic identification of the sensor upon connection and ensures that the most recent calibration is used for future measurements.

Flexible calibration
For calibration you can choose between 13 pre-defined standards, set-up a user-defined standard or enter the cell constant manually in case you know its exact value.

Connectivity and peripherals
USB and RS232 interfaces allow you to connect various peripherals from METTLER TOLEDO simultaneously, increasing the possibilities of your instrument.

Legal Information

ISM is a registered trademark of Mettler-Toledo, Inc.
InLab is a registered trademark of Mettler-Toledo, Inc.
SevenCompact is a trademark of Mettler-Toledo, Inc.
uPlace is a trademark of Mettler-Toledo, Inc.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service