Key Documents
MT11124303/UK
Mettler Toledo Printer RS-P26, RS232 interface
AC/DC input 230 V AC, UK plug
Select a Size
1 EA
CHF 1’020.00
CHF 1’020.00
Check Cart for Availability
Select a Size
Change View
1 EA
CHF 1’020.00
About This Item
UNSPSC Code:
41115600
NACRES:
NB.61
CHF 1’020.00
Check Cart for Availability
feature
GxP compliant
manufacturer/tradename
Mettler-Toledo 11124303/UK
General description
The RS-P26 Compact Printer generates GxP-compliant printouts on regular or adhesive paper. The ergonomic design makes operation easy and allows tarring and printing of the balance directly from the printer. Thanks to the compact footprint, the RS-P26 printer finds place even on smallest work places.
Choose from one of the most recent versions:
Certificates of Analysis (COA)