M29703
Methyl 2-aminobenzoate
99%
Synonym(s):
Methyl anthranilate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
vapor pressure
1 mmHg ( 20 °C)
Assay
99%
form
liquid
autoignition temp.
986 °F
refractive index
n20/D 1.582 (lit.)
bp
256 °C (lit.)
mp
24 °C (lit.)
density
1.168 g/mL at 25 °C (lit.)
SMILES string
COC(=O)c1ccccc1N
InChI
1S/C8H9NO2/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5H,9H2,1H3
InChI key
VAMXMNNIEUEQDV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
262.4 °F - closed cup
Flash Point(C)
128 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Physiology & behavior, 62(6), 1299-1302 (1998-01-07)
The relative importance of taste and odour cues in a one-trial passive avoidance learning (PAL) task was examined. One-day-old chicks were presented with a small bead and different combinations of the taste and odour of methyl anthranilate (MeA). The chicks
Angewandte Chemie (International ed. in English), 48(10), 1830-1833 (2009-01-27)
Pd and CO--ureally got me! The title reaction proceeds efficiently at 18 degrees C under CO (1 atm) with 5 % [Pd(OTs)(2)(MeCN)(2)] as precatalyst. Depending on the solvents used, either anthranilates or cyclic imides can be obtained in high yields
Carbohydrate research, 344(18), 2606-2608 (2009-11-03)
A 2:1 complex between cyclomaltoheptaose (beta-cyclodextrin) and N-methylanthranilic acid has been studied in the solid state. The inclusion complex belongs to the triclinic system (space group P1) with unit cell dimensions a=15.2773(15)A, b=15.4710(15)A, c=17.9627(18)A, alpha=99.632(5) degrees , beta=113.416(5) degrees
Journal of agricultural and food chemistry, 53(24), 9586-9589 (2005-11-24)
Bacillus megaterium, isolated in a screening process from topsoil, was used for N-demethylation of natural N-methyl methyl anthranilate to produce natural methyl anthranilate. Maximal productivity of 70 mg/L/day was achieved under laboratory-scale conditions without further optimization. No byproducts were observed.
Pest management science, 57(3), 248-252 (2001-07-18)
Two methyl anthranilate formulations, ReJex-iT TP-40 and AG-36, were tested as bird-repelling agents for animal feed and vegetable sprouts, respectively. Feral pigeons (Columbia livia) and house sparrows (Passer domesticus) are important pests of animal husbandry and field crops in Israel.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service