Skip to Content
Merck
All Photos(1)

Key Documents

LR40000-HH

Sigma-Aldrich

Cap 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106

type

for DNA synthesis

Quality Level

product line

Proligo Reagents

form

liquid

packaging

pkg of 1 × 2.5 L (LR40250-01)

impurities

≤200 ppm water content (Karl Fischer)

Preparation Note

Acetonitrile / Lutidine / N-Methylimidazole 8/1/1 (v/v/v)

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service