Skip to Content
Merck
All Photos(1)

Key Documents

JKA7216

Sigma-Aldrich

4arm-PEG10K

average MN 10,000, Dibenzocyclooctyne

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51171641
NACRES:
NA.23
Pricing and availability is not currently available.

Product Name

4-Arm PEG10K - Dibenzocyclooctyne,

form

solid

storage temp.

−20°C

Application

Activated PEG derivatives can be used to modify peptides, proteins, or in other bioconjugation applications. PEGylated materials have found broad use in drug delivery systems, virology, and immunology, as the incorporation of PEG improves pharmacological properties such as increased water solubility, enhanced resistance to degradation (protein hydrolysis), increased circulation half-life, and reduced antigenicity. In addition to PEGylation, activated PEG derivatives can also be used to form networks for tissue engineering or drug delivery applications, depending on the architecture and reactivity.

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service