Skip to Content
Merck
All Photos(2)

Key Documents

GF80594196

Molybdenum

powder, mean particle size (fsss) <5 micron, weight 500 g, purity 99.9+%

Synonym(s):

MO006021

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CHF 441.00

CHF 441.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
CHF 441.00

About This Item

Empirical Formula (Hill Notation):
Mo
CAS Number:
Molecular Weight:
95.94
MDL number:
UNSPSC Code:
12141727
PubChem Substance ID:
NACRES:
NA.23

CHF 441.00


Check Cart for Availability

Request a Bulk Order

Assay

≥99.9%

form

powder

manufacturer/tradename

Goodfellow 805-941-96

resistivity

5.0 μΩ-cm, 20°C

weight

500 g

mean particle size

<5 μm (FSSR)

bp

4612 °C (lit.)

mp

2617 °C (lit.)

density

10.3 g/mL at 25 °C (lit.)

SMILES string

[Mo]

InChI

1S/Mo

InChI key

ZOKXTWBITQBERF-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Repr. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service