Skip to Content
Merck
All Photos(1)

Documents

GF76930162

Stainless Steel - AISI 430 alloy, Fe81Cr17MnSiCSP

foil, 50 x 50mm, 0.15mm thickness, annealed

Synonym(s):

FF230280

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11171600
NACRES:
NA.23

form

foil (annealed)

manufacturer/tradename

Goodfellow 769-301-62

size × thickness

50 mm × 50 mm × 0.15 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service