Skip to Content
Merck
All Photos(1)

Key Documents

GF41696781

Iron oxide

powder, max. particle size 50 micron, weight 200 g, purity 99.9%

Synonym(s):

FE596010

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CHF 627.00

CHF 627.00


Estimated to ship on01 April 2025


Request a Bulk Order

Select a Size

Change View
1 EA
CHF 627.00

About This Item

UNSPSC Code:
12352303
NACRES:
NA.23

CHF 627.00


Estimated to ship on01 April 2025


Request a Bulk Order

Assay

99.9%

form

powder

manufacturer/tradename

Goodfellow 416-967-81

weight

200 g

max. part. size

50 μm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service