GF22793373
Erbium
powder, max. particle size 500 micron, weight 10 g, purity 99.9%
Synonym(s):
ER006010
About This Item
Assay
99.9%
form
powder
manufacturer/tradename
Goodfellow 227-933-73
resistivity
86 μΩ-cm, 20°C
weight
10 g
max. part. size
500 μm
bp
2868 °C (lit.)
mp
1529 °C (lit.)
density
9.062 g/mL at 25 °C (lit.)
SMILES string
[Er]
InChI
1S/Er
InChI key
UYAHIZSMUZPPFV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service