Skip to Content
Merck
All Photos(2)

Key Documents

GF05858376

Lead

foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)

Synonym(s):

Lead, PB000261

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pb
CAS Number:
Molecular Weight:
207.20
MDL number:
UNSPSC Code:
12141723
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

foil

manufacturer/tradename

Goodfellow 058-583-76

resistivity

20.65 μΩ-cm

size × thickness

25x25 mm × 0.05 mm

bp

1740 °C (lit.)

mp

327.4 °C (lit.)

SMILES string

[Pb]

InChI

1S/Pb

InChI key

WABPQHHGFIMREM-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1A - STOT RE 1 Oral

Target Organs

Central nervous system,Blood,Immune system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steven D Fallon et al.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons, 64(2), 223-229 (2006-01-18)
The purpose of this study was to determine if condylar morphology can be accurately depicted on the panoramic radiograph and if variations in condylar anatomy affect the radiographic appearance of the condyle. The shape and angulation of the condyle were
Leonard J S Tsuji et al.
Journal (Canadian Dental Association), 71(1), 35-38 (2005-01-15)
The lead content of the foil backing of 4 types of intraoral film commonly used by dentists was 69% to 85%. An environmental issue exists because these foils are typically thrown out with regular refuse, even though recycling programs exist.
Adam S C Siu et al.
Implant dentistry, 12(1), 35-40 (2003-04-23)
Preoperative radiographic assessment is mandatory for the meticulous treatment planning and successful placement of implants. The precise topographical evaluation of selected implant sites, apart from the most straightforward cases, requires tomographic examination with the use of diagnostic radiographic templates. The
Pardeep K Bansal et al.
Contemporary clinical dentistry, 2(2), 76-78 (2011-10-01)
The need for denture labeling is important for forensic and social reasons in case patients need to be identified individually. The importance of denture marking has long been acknowledged by the dental profession. Over the years, various denture marking systems
Karina Lopes Devito et al.
Journal of applied oral science : revista FOB, 12(4), 290-293 (2004-12-01)
All materials added to teeth should present an adequate radiopacity to allow the detection of secondary caries. Usually, in extensive cavities, base materials like calcium hydroxide cement are used for the purpose of protecting the pulp. In an attempt to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service