Skip to Content
Merck
All Photos(2)

Key Documents

E28501

Sigma-Aldrich

Ethyl 2-furoate

99%

Synonym(s):

Ethyl furan-2-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O3
CAS Number:
Molecular Weight:
140.14
Beilstein:
2653
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

bp

196 °C (lit.)

mp

32-37 °C (lit.)

density

1.117 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)c1ccco1

InChI

1S/C7H8O3/c1-2-9-7(8)6-4-3-5-10-6/h3-5H,2H2,1H3

InChI key

NHXSTXWKZVAVOQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Majdi Abid et al.
Designed monomers and polymers, 23(1), 16-24 (2020-03-05)
A series of novel-sulfonated furanic polyesteramides have been synthesized by polycondensation in the melt-phase. The copolymers were obtained with reasonable molecular weights and high inherent viscosities between 0.2 and 0.4 dL/g. The hydrolytic degradability of polyesteramides was enhanced by the
Nadhem Aissani et al.
Journal of agricultural and food chemistry, 63(27), 6120-6125 (2015-06-18)
Research on new pesticides based on plant extracts, aimed at the development of nontoxic formulates, has recently gained increased interest. This study investigated the use of the volatilome of rucola (Eruca sativa) as a powerful natural nematicidal agent against the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service