Skip to Content
Merck
All Photos(1)

Documents

DWKBPB1000P

Azlon Tapered Beakers with Printed Graduations

polypropylene, capacity 1000 mL

Synonym(s):

BPB1000P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121803
NACRES:
NB.43

material

polypropylene

packaging

pkg of 5 ea

manufacturer/tradename

Azlon

storage condition

room temperature

assay range

accuracy: 100 mL

capacity

1000 mL

Looking for similar products? Visit Product Comparison Guide

Application

Excellent solution for measuring and dispensing laboratory solutions and reagents

Features and Benefits

Autoclavable at 121°C and complies with ISO 7056

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service