Skip to Content
Merck
All Photos(1)

Documents

DWK986701

WHEATON® liquid scintillation vial with attached foil lined PP cap, glass

lips on vial transparent high-density polyethylene bottle, capacity (20 mL), screw cap

Synonym(s):

986701

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

material

lips on vial transparent high-density polyethylene bottle
metal foil liner
white polypropylene cap

feature

with cap

packaging

case of 500 ea individual cells in trays of 100

manufacturer/tradename

WHEATON® 986701

bottle diam. × H × W

(27 mm) (61 mm) (302 mm)

capacity

(20 mL)

fitting

22-400 neck
22-400

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • High-density polyethylene bottle made of low potassium borosilicate glass or polyethylene terephthalate or high-density polyethylene contains white polypropylene screw cap
  • Background counts are consistent and low; ultraviolet transmission is high
  • Available with cap attached, caps packed separately or without cap

Legal Information

WHEATON is a registered trademark of Wheaton Industries, Inc.

related product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service