DWK986586
WHEATON® liquid scintillation vial with seperate PE cone lined urea cap
lips on vial transparent borosilicate glass bottle, capacity (20 mL), screw cap
Synonym(s):
986586, Scintillation Vial with Cap, WHEATON Scintillation Vial
About This Item
Recommended Products
material
lips on vial transparent borosilicate glass bottle
white cap
sterility
non-sterile
feature
with cap
packaging
case of 500 ea Vials shrink-wrapped trays Screw caps in a separate sealed tray
manufacturer/tradename
WHEATON® 986586
bottle diam. × H × W
(28 mm) (61 mm) (387 mm)
capacity
(20 mL)
fitting
22-400 neck
22-400
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
- Vial offers low permeability to solvents
- Minimal background counts
- Vials can be incinerated without emission of harmful gases
- Comes along with utility trays that enable the storage of samples
- Made of WHEATON® 180 low potassium clear type 1A borosilicate glass
Legal Information
related product
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service