Skip to Content
Merck
All Photos(1)

Documents

DWK374322

Wheaton® Hose Barb / Luer Adapter For Attaching Manifold (Fits 2 And 3Mm)

Synonym(s):

374322

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815
NACRES:
NB.43

Sterilization Compatibility

autoclavable compatible

packaging

pkg of 1 ea

manufacturer/tradename

WHEATON® 374322

Application

Research, Development and Quality Control, Pharmaceutical and Biotechnology, Food and Beverage, Waste Water and Sample Collection

Features and Benefits

  • The luer adapter allows connection to equipment and components with a female luer connector

Legal Information

WHEATON is a registered trademark of Wheaton Industries, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service