DWK218663653
DURAN® wide mouth amber laboratory bottle
with cap, bottle capacity (250 mL), round amber glass bottle, non-sterile
Synonym(s):
218663653
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
material
round amber glass bottle
sterility
non-sterile
feature
with cap
packaging
case of 10 ea
manufacturer/tradename
DURAN® 218663653
bottle capacity
(250 mL)
fitting
GL 80 cap
GL 80 neck
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
UV protection up to approx. 500 nm due to external colouration. The only external application of the colour ensures that internal surfaces have standard DURAN glass properties. Special thread design for rapid opening and closing in less than a turn Optimum filling and pouring and particularly easy cleaning. Spoons, spatulas and tweezers can be manipulated with ease.
Even large-volume funnels can be inserted into the bottles without any difficulty. Stability due to the large base areaEasy-to-read scale with graduation marks to estimate the contentFired-on, highly durable white ceramic print.
Even large-volume funnels can be inserted into the bottles without any difficulty. Stability due to the large base areaEasy-to-read scale with graduation marks to estimate the contentFired-on, highly durable white ceramic print.
Features and Benefits
- Manufactured from ambered DURAN® borosilicate glass 3.3. DURAN® is a neutral glass, classified as a Glass Type I according to USP <660>, EP (3.2.1) and JP <7.01>, and meets ASTM E438, Type I, Class A requirements
- USP <660> (Spectral Transmission) and EP (3.2.1) compliant. Less than 10% spectral transmission between 290 nm and 450 nm.
- Easy access to the bottle contents due to the wide diameter of the bottle neck
- Special thread design for rapid opening and closing in less than a turn
- Optimum filling and pouring and particularly easy cleaning
- Spoons, spatulas and tweezers can be manipulated with ease
- Even large-volume funnels can be inserted into the bottles without any difficulty
- Stability due to the large base area
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service