Skip to Content
Merck
All Photos(1)

Documents

D54850

Sigma-Aldrich

2,2-Dichloroacetophenone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCHCl2
CAS Number:
Molecular Weight:
189.04
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid

refractive index

n20/D 1.5686 (lit.)

bp

132-134 °C/13 mmHg (lit.)

mp

20-21 °C (lit.)

density

1.34 g/mL at 25 °C (lit.)

SMILES string

ClC(Cl)C(C1=CC=CC=C1)=O

InChI

1S/C8H6Cl2O/c9-8(10)7(11)6-4-2-1-3-5-6/h1-5,8H

InChI key

CERJZAHSUZVMCH-UHFFFAOYSA-N

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Swatishree Sradhanjali et al.
PloS one, 12(5), e0177744-e0177744 (2017-05-16)
Pyruvate dehydrogenase kinase 1 (PDK1), a key enzyme implicated in metabolic reprogramming of tumors, is induced in several tumors including glioblastoma, breast cancer and melanoma. However, the role played by PDK1 is not studied in retinoblastoma (RB). In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service