Skip to Content
Merck
All Photos(1)

Key Documents

D189790

Sigma-Aldrich

5,7-Dimethyl-s-triazolo[1,5-a]pyrimidine

98%

Synonym(s):

5,7-Dimethyl-1,3,4-triazaindolizine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8N4
CAS Number:
Molecular Weight:
148.17
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

98%

form

solid

mp

136-139 °C (lit.)

SMILES string

Cc1cc(C)n2ncnc2n1

InChI

1S/C7H8N4/c1-5-3-6(2)11-7(10-5)8-4-9-11/h3-4H,1-2H3

InChI key

WXVQCFALDUVKSC-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Hering et al.
Biomedica biochimica acta, 44(5), K37-K41 (1985-01-01)
The action of trapidil (RocornalR) and its derivatives AR 12-456 and AR 12-160 on the inward calcium current (ICa) was studied in mouse neuroblastoma x rat glioma hybrid cells of the line 108CC5 under voltage clamp conditions by means of
J M Salas et al.
Journal of inorganic biochemistry, 64(4), 259-271 (1996-12-01)
The preparation and spectroscopic characterization of a series of dmtp complexes of Zn, Cd, and Hg, where dmtp is 5,7-dimethyl[1,2,4]triazolo[1,5-a]pyrimidine, is reported. Crystal structure analyses of [Zn(dmtp)2Br2] and [Hg(dmtp)2Cl2] reveal distorted tetrahedral geometries about the central atoms and the dmtp
Marzena Fandzloch et al.
Journal of inorganic biochemistry, 210, 111072-111072 (2020-06-21)
Five novel rhodium(II) complexes of general formula [Rh2(μ-OOCCH3)4L2], where L is a triazolopyrimidine derivative, in particular dimethyl-1,2,4-triazolo[1,5-a]pyrimidine (dmtp) for (1), 5,7-diethyl-1,2,4-triazolo[1,5-a]pyrimidine (detp) for (2), 7-isobutyl-5-methyl-1,2,4-triazolo[1,5-a]pyrimidine (ibmtp) for (3), 7-hydroxy-5-methyl-1,2,4-triazolo[1,5-a]pyrimidine (HmtpO) for (4) and 5,7-ditertbutyl-1,2,4-triazolo[1,5-a]pyrimidine (dbtp) for (5) are reported. These

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service