Key Documents
CLS9998241
Corning® phenolic cap, PTFE liner
24-410 thread
Synonym(s):
black phenolic, phenolic caps, screw caps, Corning® 9998
Select a Size
144 EA
CHF 328.00
CHF 328.00
Check Cart for Availability
Select a Size
Change View
144 EA
CHF 328.00
About This Item
UNSPSC Code:
41122500
NACRES:
NB.43
CHF 328.00
Check Cart for Availability
material
PTFE liner
black phenolic cap
manufacturer/tradename
Corning 9998-241
H
~18.5 mm
fitting
24-410 thread
General description
These reusable Corning caps are fabricated from specially formulated resin to resist the effects of temperature and steam in autoclaving. Caps are provided with a glued-in rubber liner over which is firmly bonded a PTFE interface to provide a highly inert sealing face.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)