CLS980012
Pyrex® test tubes, beaded rim
capacity 5 mL
Synonym(s):
Corning® 9800
About This Item
Recommended Products
material
borosilicate glass
plain neck
round bottom tube
sterility
non-sterile
feature
graduations
packaging
pack of 72
case of 720
manufacturer/tradename
Corning 9800-12
O.D. × L
12 mm × 75 mm
capacity
5 mL
color
clear
suitability
suitable for (test)
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service