CLS808015
Pyrex® centrifuge tubes with beaded rim and white enamel graduations
capacity 15 mL
Synonym(s):
Corning® 8080
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
borosilicate glass
conical bottom tube
sterility
non-sterile
feature
cap: no
packaging
case of 12
pack of 12
manufacturer/tradename
Corning 8080-15
O.D. × L
17 mm × 120 mm
capacity
15 mL
color
clear
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Formed from borosilicate glass. Stress that glass can withstand is greatly decreased by surface flaws such as scratches, chips, and cracks. Always use with correct sleeve or holder.
Other Notes
15 mL sizes have 0.1 mL graduations upward from tip; 50 mL size has 0.5 mL graduations upward from tip to 10 mL and 1 mL graduations from 10 to 50 mL.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service