Skip to Content
Merck
All Photos(1)

Documents

CLS7103C5

Pyrex® volumetric pipette, certified, to contain, to deliver, Class A

volume 5 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.43

material

glass

feature

class A
graduation

packaging

pack of 2 ea
case of 6 ea

manufacturer/tradename

Corning 7103C-5

volume measuring range

5 mL

capacity

5 mL

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Pyrex® volumetric pipettes (volume 5 mL) are calibrated to Class A tolerances in accordance with ASTM E-542 and ASTM E-969.
  • Each pipette is individually serialized and supplied with a Certificate of Identification and Capacity, traceable to NIST standards.
  • These precision PYREX® volumetric pipettes are manufactured and calibrated "To Contain" (TC - bottom line) and "To Deliver" (TD - top line) to specifications for Class A volumetric ware.
  • Color-coded markings are enameled onto the glass.
  • Tested and calibrated in an ISO/IEC 17025 certified laboratory.

Legal Information

PYREX is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service