Key Documents
CLS4965B500
Pyrex® three-neck round-bottom flask
capacity 500 mL, center joint: ST/NS 19/22, side joint: ST/NS 19/22
Synonym(s):
Pyrex® round-bottom three neck angle type distilling flask, bottom flask, flask round, flask round bottom
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Pricing and availability is not currently available.
material
borosilicate glass
round bottom flask
three-neck (Vertical)
Joint
center joint: ST/NS19/22
side joint: ST/NS19/22
sterility
non-sterile
feature
closure type Standard Taper joint
packaging
case of 1
manufacturer/tradename
Corning 4965B-500
capacity
500 mL
neck size
(3)
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)