Skip to Content
Merck
All Photos(1)

Key Documents

CDS014167

Sigma-Aldrich

2-ethyl-3,5(6)-dimethyl-pyrazine

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2
CAS Number:
Molecular Weight:
136.19
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

CCc1ncc(C)nc1C

InChI

1S/C8H12N2/c1-4-8-7(3)10-6(2)5-9-8/h5H,4H2,1-3H3

InChI key

JZBCTZLGKSYRSF-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenneth Smith et al.
Molecules (Basel, Switzerland), 25(9) (2020-05-15)
Differences in the aroma profiles of extruded maize puffs made from refined grain and whole grain flour were investigated. Gas chromatography/mass spectrometry/olfactometry (GC/MS/O) analysis reported 13 aroma compounds with a flavor dilution (FD) value ≥16. Quantitative analysis identified eight compounds
Ahmet Salih Sonmezdag et al.
Food chemistry, 240, 24-31 (2017-09-28)
Volatile, aroma-active, and phenolic compounds of pistachio oil obtained from cv. Uzun were investigated in the current study. To obtain a representative aromatic extract, three of the most widely used extraction methods were compared using a representative test; the solvent-assisted
Julien Brechbühl et al.
Communications biology, 3(1), 533-533 (2020-09-26)
The ability to efficiently search for food is fundamental for animal survival. Olfactory messages are used to find food while being aware of the impending risk of predation. How these different olfactory clues are combined to optimize decision-making concerning food
Aileen Pua et al.
Food chemistry, 302, 125370-125370 (2019-08-24)
Four Arabica coffees (Brazil, Colombia, Ethiopia, and Guatemala) yield highly variant odours, attesting to the complexities of coffee aroma that command advanced analytical tools. In this study, their volatiles were extracted using solvent-assisted flavour evaporation (SAFE) and headspace solid-phase microextraction
Mohamed A A Mahmoud et al.
Frontiers in chemistry, 6, 241-241 (2018-07-11)
Although the microbiota is considered to be the primary source of off-flavors in farmed fish, there is a lack of information about the possible contribution of feeds to fish malodor. For this reason, the current study was designed to perform

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service