Skip to Content
Merck
All Photos(1)

Key Documents

CDS001219

Sigma-Aldrich

3-Hepten-2-one

AldrichCPR

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
CHF 84.60

CHF 84.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
CHF 84.60

About This Item

Empirical Formula (Hill Notation):
C7H12O
CAS Number:
Molecular Weight:
112.17
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:

CHF 84.60


Check Cart for Availability

Request a Bulk Order

form

liquid

SMILES string

CCC\C=C\C(C)=O

InChI

1S/C7H12O/c1-3-4-5-6-7(2)8/h5-6H,3-4H2,1-2H3/b6-5+

InChI key

JHHZQADGLDKIPM-AATRIKPKSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.2 °F

Flash Point(C)

49 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hilaire Kpongbe et al.
Journal of chemical ecology, 45(7), 570-578 (2019-06-19)
The brown spiny bug, Clavigralla tomentosicollis Stål (Hemiptera: Coreidae) is a key pest of leguminous crops in many countries in Africa, causing significant yield losses especially in cowpea, pigeon pea and common beans. Although C. tomentosicollis uses olfaction to aggregate
Svenja Nörenberg et al.
Journal of agricultural and food chemistry, 65(40), 8913-8922 (2017-09-19)
A homologous series of 4-mercapto-2-alkanols (C5-C10) was used to investigate the impact of the stereochemistry on the sensory properties of a class of naturally occurring polyfunctional thiols having a 1,3-oxygen-sulfur functionality. Stereoisomers were obtained via syntheses of racemic mixtures and
Akiko Fujita et al.
PloS one, 15(2), e0229269-e0229269 (2020-02-23)
Psychiatric disorders including depression and anxiety comprise a broad range of conditions with different symptoms. We have developed a mouse model of depression/anxiety in mice deficient in the St3gal4 gene. In this study, we performed a comparative analysis of urinary

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service