CAD4052
Standard point hypodermic needles, 304 stainless steel
gauge 21, L 1 1/2 in.
All Photos(1)
About This Item
UNSPSC Code:
42142523
NACRES:
NB.03
material
304 stainless steel needle
packaging
pack of 12 ea
manufacturer/tradename
Cadence Science 4052
L
1 1/2 in.
gauge
21
General description
Hypodermic needles are precision made and designed to ensure strict adherence with U.S. Government specification GG-N-196. Points are machine ground to achieve uniform sharpness. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure freedom from burrs and debris. All needle cannula are fabricated of type 304 stainless steel tubing which must pass rigorous quality control prior to acceptance and use.
Standard nickle plated Luer hubs comply with Luer taper specifications ISO 594. This assures an interchangeable fit mating with Luer slips and Luer locks. Contoured hub cavity facilitates cleaning.
Standard nickle plated Luer hubs comply with Luer taper specifications ISO 594. This assures an interchangeable fit mating with Luer slips and Luer locks. Contoured hub cavity facilitates cleaning.
Choose from one of the most recent versions:
Certificates of Analysis (COA)