Skip to Content
Merck
All Photos(1)

Key Documents

C94408

Sigma-Aldrich

4-Cyano-4-phenylpiperidine hydrochloride

96%

Synonym(s):

4-Phenyl-4-piperidinecarbonitrile hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
CHF 148.00

CHF 148.00


Estimated to ship on13 April 2025


Request a Bulk Order

Select a Size

Change View
10 G
CHF 148.00

About This Item

Empirical Formula (Hill Notation):
C12H14N2 · HCl
CAS Number:
Molecular Weight:
222.71
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

CHF 148.00


Estimated to ship on13 April 2025


Request a Bulk Order

Quality Level

Assay

96%

form

solid

mp

208-210 °C (lit.)

SMILES string

Cl.N#CC1(CCNCC1)c2ccccc2

InChI

1S/C12H14N2.ClH/c13-10-12(6-8-14-9-7-12)11-4-2-1-3-5-11;/h1-5,14H,6-9H2;1H

InChI key

CQPHZBOPSZGTJM-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • CCR5 antagonists derived at the N-terminal of the piperidine ring
  • Substituted piperidines as lignads of chemokine receptor 5 and inhibitors of HIV-1
  • Functionalized nitrogen heterocycles by radical decarboxylation of β- and γ- amino acids
  • Benzovesamicol analogs as potential imaging probes for vesicular acetylcholine transporters
  • Neurotransmitter transporter inhibitors with activity at dopamine receptor sites
  • α1a Adrenoceptor-selective antagonists

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service