C4265
p-Chloro-β-methylphenethylamine hydrochloride
≥98%
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Quality Level
Assay
≥98%
form
powder
storage temp.
2-8°C
SMILES string
Cl[H].CC(CN)c1ccc(Cl)cc1
InChI
1S/C9H12ClN.ClH/c1-7(6-11)8-2-4-9(10)5-3-8;/h2-5,7H,6,11H2,1H3;1H
InChI key
RTPMRVSAKBTCBG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Neuropharmacology, 30(4), 329-335 (1991-04-01)
Various intraperitoneal doses of 5-fluoro-alpha-methyltryptamine (5-FMT), given to mice, dose-dependently inhibited only MAOA activity, with similar degrees of inhibition in the striatum, hypothalamus and the rest of the forebrain. The activity inhibited in these regions, completely recovered to control levels
Research communications in chemical pathology and pharmacology, 54(1), 125-128 (1986-10-01)
In order to develop new and potent reversible MAO-B-selective inhibitors, we examined the inhibitory effects of two substrate-analogues, 5-fluoro-alpha-methyltryptamine (5-FMT) and p-chloro-beta-methylphenethylamine (p-CMP) on rat brain MAO-A and -B activity in vitro. Studies of the inhibitory effects of both analogues
Japanese journal of pharmacology, 46(2), 197-199 (1988-02-01)
To further clarify highly MAO-A- or -B-selective inhibitory properties of 5-fluoro-alpha-methyltryptamine (5-FMT) and p-chloro-beta-methylphenethylamine (p-CMPEA), we determined the types and K1 values of inhibition of rat brain MAO-A and -B activity in vitro. The kinetic data obtained showed that 5-FMT
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service