Skip to Content
Merck
All Photos(1)

Key Documents

BR781686

BRAND® microplate BRANDplates®, pureGrade

size 384 wells, size 120 μL, flat bottom, white, with transparent bottom

Synonym(s):

BRANDplates®, Microplates, pureGradeS

Sign Into View Organizational & Contract Pricing

Select a Size

50 EA
CHF 846.00

CHF 846.00


Estimated to ship on11 May 2025



Select a Size

Change View
50 EA
CHF 846.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
flat bottom
polystyrene
white wells
size:
120 μL
384 wells
working volume:
120 μL

CHF 846.00


Estimated to ship on11 May 2025


material

flat bottom
polystyrene
white wells

sterility

sterile

feature

lid
skirt (F-bottom, transparent bottom)

packaging

pack of 50 ea
pkg of (individually packaged)

manufacturer/tradename

BRAND 781686

size

120 μL
384 wells

working volume

120 μL

color

white, with transparent bottom

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

The BRAND® Microplate BRANDplates® PureGrade features a non-treated surface, making it particularly well-suited for bacteriological assays. Sterile according to ISO 11137 standards, this microplate ensures a contamination-free environment for sensitive experiments. It is classified as BIO-CERT® CELL CULTURE QUALITY, guaranteeing that it is free from human DNA, RNase, DNase, and pyrogens, and is non-cytotoxic. This reliable microplate is an essential tool for researchers conducting microbiological studies, providing consistent performance and safety in cell culture applications.

Legal Information

BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service